The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033402.4 | NP_208325.3 | 14 - 15 | 2538 | 115 |
XM_005251318.3 | XP_005251375.1 | 13 - 14 | 2327 | 115 | |
XM_005251319.4 | XP_005251376.1 | 12 - 13 | 2021 | 115 | |
XM_005251320.3 | XP_005251377.1 | 11 - 12 | 1813 | 115 | |
XM_005251322.3 | XP_005251379.1 | 10 - 11 | 1627 | 115 | |
XM_017013920.1 | XP_016869409.1 | 15 - 16 | 2486 | 115 | |
XM_017013921.1 | XP_016869410.1 | 15 - 16 | 2423 | 115 | |
XM_017013922.1 | XP_016869411.1 | 14 - 15 | 2300 | 115 | |
XM_017013923.1 | XP_016869412.1 | 14 - 15 | 2313 | 115 | |
XM_017013925.1 | XP_016869414.1 | 12 - 13 | 2032 | 115 | |
XM_017013926.1 | XP_016869415.1 | 12 - 13 | 2015 | 115 | |
XM_017013927.1 | XP_016869416.1 | 13 - 14 | 2043 | 115 | |
XM_017013928.1 | XP_016869417.1 | 10 - 11 | 1495 | 115 | |
GenBank mRNA | AB051551.1 | - | 13 - 14 | 3159 | 115 |
AK056185.1 | - | 13 - 14 | 2501 | 115 | |
AK057990.1 | - | 5 - 6 | 793 | 115 | |
AK300874.1 | - | 13 - 14 | 2182 | 115 | |
AK302648.1 | - | 14 - 15 | 2388 | 115 | |
AK307443.1 | - | 11 - 12 | 1794 | 115 | |
AK307636.1 | - | 13 - 14 | 2172 | 115 | |
BC030701.2 | - | 14 - 15 | 2411 | 115 |