Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX16
Entrez Gene ID
64089
Gene Name
sorting nexin 16
Gene Aliases
-
Chromosome Location
Chr.8: 81799583 - 81842326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022133.3 NP_071416.2 5 - 6 1116 115
NM_152836.2 NP_690049.1 4 - 5 870 115
NM_152837.2 NP_690050.1 3 - 4 783 115
XM_005251282.4 XP_005251339.1 5 - 6 823 115
XM_005251283.2 XP_005251340.1 6 - 7 960 115
XM_011517574.2 XP_011515876.1 3 - 4 213 115
GenBank mRNA AF305779.1 - 4 - 5 721 115
AK290903.1 - 4 - 5 786 115
AL833763.1 - 3 - 4 690 115
BC033630.1 - 4 - 5 778 115

View Full Product Details