The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169154.1 | NP_001162625.1 | 2 - 3 | 330 | 160 |
XM_011530855.2 | XP_011529157.1 | 2 - 3 | 170 | 160 | |
GenBank mRNA | AI147427.1 | - | 1 - 2 | 286 | 160 |