Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D3
Entrez Gene ID
139212
Gene Name
PIH1 domain containing 3
Gene Aliases
CXorf41, NYSAR97
Chromosome Location
Chr.X: 107206629 - 107244243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169154.1 NP_001162625.1 4 - 5 559 81
NM_173494.1 NP_775765.1 3 - 4 284 81
XM_011530855.2 XP_011529157.1 4 - 5 399 81
GenBank mRNA AY211928.1 - 3 - 4 279 81
BC033510.1 - 3 - 4 284 81

View Full Product Details