The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169154.1 | NP_001162625.1 | 6 - 7 | 764 | 156 |
NM_173494.1 | NP_775765.1 | 5 - 6 | 489 | 156 | |
XM_011530855.2 | XP_011529157.1 | 6 - 7 | 604 | 156 | |
GenBank mRNA | AY211928.1 | - | 5 - 6 | 484 | 157 |
BC033510.1 | - | 5 - 6 | 489 | 156 |