The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015346.3 | NP_056161.2 | 20 - 21 | 3768 | 87 |
XM_011536609.2 | XP_011534911.1 | 20 - 21 | 3769 | 87 | |
XM_017021124.1 | XP_016876613.1 | 20 - 21 | 3769 | 87 | |
XM_017021125.1 | XP_016876614.1 | 20 - 21 | 3769 | 87 | |
XM_017021126.1 | XP_016876615.1 | 19 - 20 | 4383 | 87 | |
XM_017021127.1 | XP_016876616.1 | 9 - 10 | 1465 | 87 | |
XM_017021128.1 | XP_016876617.1 | 9 - 10 | 1318 | 87 | |
GenBank mRNA | AB002319.1 | - | 5 - 6 | 638 | 87 |
AB425197.1 | - | 20 - 21 | 3756 | 87 | |
BC144594.1 | - | 18 - 19 | 3391 | 87 | |
BX537886.1 | - | 20 - 21 | 3762 | 87 | |
BX538025.1 | - | 16 - 17 | 3261 | 87 | |
BX648683.1 | - | 20 - 21 | 3766 | 87 | |
CR749276.1 | - | 21 - 22 | 3703 | 87 | |
CR749365.1 | - | 20 - 21 | 3767 | 87 |