Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMND1
Entrez Gene ID
55005
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3
Chromosome Location
Chr.6: 151404548 - 151452181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017909.3 NP_060379.2 2 - 3 689 73
XM_017010988.1 XP_016866477.1 2 - 3 662 73
GenBank mRNA AK292339.1 - 2 - 3 664 73
BC012081.1 - 1 - 2 526 73
BC106065.1 - 2 - 3 651 73
BC119683.1 - 2 - 3 554 73

View Full Product Details