The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282704.1 | NP_001269633.1 | 7 - 8 | 944 | 150 |
NM_001282705.1 | NP_001269634.1 | 6 - 7 | 966 | 150 | |
NM_001282706.1 | NP_001269635.1 | 6 - 7 | 796 | 150 | |
NM_001282707.1 | NP_001269636.1 | 6 - 7 | 771 | 150 | |
NM_005742.3 | NP_005733.1 | 5 - 6 | 593 | 150 | |
XM_011510308.1 | XP_011508610.1 | 5 - 6 | 534 | 150 | |
GenBank mRNA | AK127433.1 | - | 7 - 8 | 944 | 150 |
AK131234.1 | - | 6 - 7 | 962 | 150 | |
AK225496.1 | - | 4 - 5 | 339 | 150 | |
AK289428.1 | - | 5 - 6 | 469 | 150 | |
AK294347.1 | - | 6 - 7 | 686 | 150 | |
AK297547.1 | - | 6 - 7 | 756 | 150 | |
BC001312.1 | - | 5 - 6 | 549 | 150 | |
D49489.1 | - | - | 544 | 150 | |
U79278.1 | - | 4 - 5 | 395 | 150 |