Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRN1
Entrez Gene ID
51299
Gene Name
neuritin 1
Gene Aliases
NRN, dJ380B8.2
Chromosome Location
Chr.6: 5997999 - 6007605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278710.1 NP_001265639.1 2 - 3 210 88
NM_016588.2 NP_057672.1 1 - 2 708 88
GenBank mRNA AF114833.1 - 1 - 2 239 88
AF136631.1 - 1 - 2 225 88
AJ420483.1 - 1 - 2 182 88
AK314095.1 - 1 - 2 261 88
AL541038.3 - 1 - 2 294 88
BC002683.2 - 1 - 2 186 88
BC042019.1 - 1 - 2 709 88
BF970432.1 - 1 - 2 101 88

View Full Product Details