The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173506.6 | NP_775777.3 | 2 - 3 | 1282 | 91 |
GenBank mRNA | AK292305.1 | - | 2 - 3 | 1277 | 91 |
AY358726.1 | - | 2 - 3 | 91 | ||
AY771680.1 | - | 1 - 2 | 68 | 91 |