The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173506.6 | NP_775777.3 | 3 - 4 | 1427 | 70 |
XM_005258543.3 | XP_005258600.1 | 3 - 4 | 455 | 70 | |
XM_005278383.4 | XP_005278440.1 | 3 - 4 | 1466 | 70 | |
XM_005278385.4 | XP_005278442.1 | 3 - 4 | 1466 | 70 | |
XM_005278387.4 | XP_005278444.1 | 3 - 4 | 1466 | 70 | |
XM_005278388.3 | XP_005278445.1 | 2 - 3 | 1250 | 70 | |
XM_006723035.2 | XP_006723098.1 | 2 - 3 | 243 | 70 | |
XM_011526483.2 | XP_011524785.1 | 3 - 4 | 623 | 70 | |
XM_011526484.2 | XP_011524786.1 | 3 - 4 | 528 | 70 | |
GenBank mRNA | AK292305.1 | - | 3 - 4 | 1422 | 70 |
AY358726.1 | - | 3 - 4 | 1196 | 70 | |
AY771680.1 | - | 2 - 3 | 213 | 70 |