The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 16 - 17 | 1874 | 133 |
NM_002647.3 | NP_002638.2 | 17 - 18 | 2063 | 133 | |
XM_011526031.2 | XP_011524333.1 | 18 - 19 | 2228 | 133 | |
GenBank mRNA | AK022653.1 | - | 1 - 2 | 72 | 133 |
AK090567.1 | - | 4 - 5 | 2597 | 133 | |
AK298517.1 | - | 16 - 17 | 1818 | 133 | |
BC033004.2 | - | 17 - 18 | 2013 | 133 | |
BC053651.1 | - | 17 - 18 | 1993 | 133 | |
Z46973.1 | - | 17 - 18 | 2016 | 133 |