The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 23 - 24 | 2548 | 64 |
NM_002647.3 | NP_002638.2 | 24 - 25 | 2737 | 64 | |
XM_011526031.2 | XP_011524333.1 | 25 - 26 | 2902 | 64 | |
GenBank mRNA | AK090567.1 | - | 11 - 12 | 3271 | 64 |
AK298517.1 | - | 23 - 24 | 2492 | 64 | |
BC033004.2 | - | 24 - 25 | 2687 | 64 | |
BC053651.1 | - | 24 - 25 | 2667 | 64 | |
CA945171.1 | - | 1 - 2 | 64 | ||
Z46973.1 | - | 24 - 25 | 2690 | 64 |