The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136139.2 | NP_001129611.1 | 17 - 18 | 1851 | 69 |
XM_006722855.3 | XP_006722918.1 | 18 - 19 | 2054 | 69 | |
XM_006722856.3 | XP_006722919.1 | 18 - 19 | 2055 | 69 | |
XM_006722857.3 | XP_006722920.1 | 18 - 19 | 2052 | 69 | |
XM_006722858.3 | XP_006722921.1 | 18 - 19 | 2053 | 69 | |
XM_011528220.2 | XP_011526522.1 | 18 - 19 | 2054 | 69 | |
XM_011528221.2 | XP_011526523.1 | 18 - 19 | 2052 | 69 | |
XM_011528225.2 | XP_011526527.1 | 18 - 19 | 1964 | 69 | |
XM_011528226.2 | XP_011526528.1 | 18 - 19 | 1964 | 69 | |
XM_011528227.2 | XP_011526529.1 | 16 - 17 | 1813 | 69 | |
XM_017027178.1 | XP_016882667.1 | 18 - 19 | 2055 | 69 | |
XM_017027179.1 | XP_016882668.1 | 18 - 19 | 2052 | 69 | |
XM_017027182.1 | XP_016882671.1 | 18 - 19 | 1966 | 69 | |
GenBank mRNA | AW028755.1 | - | 2 - 3 | 310 | 69 |
KF303591.1 | - | 15 - 16 | 1659 | 69 | |
M65214.1 | - | 2 - 3 | 282 | 69 | |
X52078.1 | - | 14 - 15 | 1605 | 69 |