The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020962.2 | NP_066013.1 | 2 - 3 | 627 | 111 |
XM_011521845.2 | XP_011520147.1 | 2 - 3 | 807 | 111 | |
XM_011521846.2 | XP_011520148.1 | 2 - 3 | 431 | 111 | |
XM_011521847.1 | XP_011520149.1 | 2 - 3 | 491 | 111 | |
XM_017022448.1 | XP_016877937.1 | 2 - 3 | 809 | 111 | |
XM_017022449.1 | XP_016877938.1 | 2 - 3 | 815 | 111 | |
GenBank mRNA | AB052622.1 | - | 2 - 3 | 627 | 111 |