The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020962.2 | NP_066013.1 | 6 - 7 | 1204 | 73 |
XM_011521845.2 | XP_011520147.1 | 6 - 7 | 1384 | 73 | |
XM_011521846.2 | XP_011520148.1 | 6 - 7 | 1008 | 73 | |
XM_011521847.1 | XP_011520149.1 | 6 - 7 | 1068 | 73 | |
XM_017022448.1 | XP_016877937.1 | 6 - 7 | 1386 | 73 | |
XM_017022449.1 | XP_016877938.1 | 6 - 7 | 1392 | 73 | |
GenBank mRNA | AB052622.1 | - | 6 - 7 | 1204 | 73 |