The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018649.2 | NP_061119.1 | 3 - 4 | 738 | 73 |
GenBank mRNA | AF151534.1 | - | 3 - 4 | 492 | 73 |
AF336304.1 | - | 3 - 4 | 470 | 73 | |
AK022475.1 | - | 3 - 4 | 462 | 73 | |
AK022776.1 | - | 3 - 4 | 493 | 73 | |
AK131057.1 | - | 3 - 4 | 489 | 73 | |
BC016172.2 | - | 3 - 4 | 494 | 73 |