The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003620.3 | NP_003611.1 | 3 - 4 | 1058 | 97 |
GenBank mRNA | AK299773.1 | - | 4 - 5 | 767 | 97 |
AK312902.1 | - | 3 - 4 | 1048 | 97 | |
BC016480.1 | - | 3 - 4 | 974 | 97 | |
BC033893.2 | - | 3 - 4 | 932 | 97 | |
BC042418.1 | - | 3 - 4 | 972 | 97 | |
BC060877.1 | - | 3 - 4 | 1058 | 97 | |
BT009780.1 | - | 3 - 4 | 826 | 97 | |
KU178420.1 | - | 3 - 4 | 826 | 97 | |
KU178421.1 | - | 2 - 3 | 597 | 97 | |
U78305.1 | - | 3 - 4 | 1029 | 97 |