The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306145.1 | NP_001293074.1 | 2 - 3 | 422 | 136 |
NM_003828.3 | NP_003819.1 | 2 - 3 | 422 | 136 | |
XM_006724855.3 | XP_006724918.1 | 2 - 3 | 422 | 136 | |
XM_011531209.2 | XP_011529511.1 | 2 - 3 | 182 | 136 | |
GenBank mRNA | AF057354.1 | - | 2 - 3 | 246 | 136 |
AJ224979.1 | - | 2 - 3 | 248 | 136 | |
BC032947.1 | - | 2 - 3 | 419 | 136 | |
DR000536.1 | - | 2 - 3 | 424 | 136 |