Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
Chromosome Location
Chr.4: 8440774 - 8512531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152544.2 NP_689757.2 1 - 2 634 111
XM_005247950.3 XP_005248007.1 1 - 2 660 111
XM_011513406.2 XP_011511708.1 1 - 2 663 111
XM_011513407.2 XP_011511709.1 1 - 2 663 111
XM_011513410.2 XP_011511712.1 1 - 2 665 111
XM_017007811.1 XP_016863300.1 1 - 2 664 111

View Full Product Details