The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152544.2 | NP_689757.2 | 2 - 3 | 749 | 87 |
XM_011513406.2 | XP_011511708.1 | 2 - 3 | 778 | 87 | |
XM_011513407.2 | XP_011511709.1 | 2 - 3 | 778 | 87 | |
XM_011513408.1 | XP_011511710.1 | 3 - 4 | 890 | 87 | |
XM_011513410.2 | XP_011511712.1 | 2 - 3 | 780 | 87 | |
XM_017007811.1 | XP_016863300.1 | 2 - 3 | 779 | 87 |