Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAALADL2
Entrez Gene ID
254827
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
-
Chromosome Location
Chr.3: 174439669 - 175809510 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006073.1 XP_016861562.1 1 - 2 137 83

View Full Product Details