The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100118.1 | NP_001093588.1 | 5 - 6 | 728 | 73 |
NM_001100119.1 | NP_001093589.1 | 6 - 7 | 822 | 73 | |
NM_005432.3 | NP_005423.1 | 6 - 7 | 785 | 73 | |
XM_005268046.2 | XP_005268103.1 | 5 - 6 | 822 | 73 | |
XM_011537138.2 | XP_011535440.1 | 6 - 7 | 768 | 73 | |
GenBank mRNA | AF035586.1 | - | 5 - 6 | 713 | 73 |
AK022829.1 | - | 6 - 7 | 714 | 73 | |
AK023646.1 | - | 6 - 7 | 758 | 73 | |
AK124498.1 | - | 4 - 5 | 1202 | 73 | |
AK126706.1 | - | 1 - 2 | 1854 | 73 | |
BC001036.1 | - | 6 - 7 | 757 | 73 | |
BC011725.2 | - | 5 - 6 | 688 | 73 | |
BT007417.1 | - | 3 - 4 | 405 | 73 | |
BX161398.1 | - | 5 - 6 | 697 | 73 |