Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KIAA0125
- Entrez Gene ID
-
9834
- Gene Name
- KIAA0125
- Gene Aliases
-
C14orf110, FAM30A, HSPC053
- Chromosome Location
-
Chr.14: 105917979 - 105932642 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026800.2 |
|
6 - 6 |
9491 |
77 |
GenBank mRNA |
AW451894.1 |
- |
1 - 1 |
|
77 |
BC152473.1 |
- |
1 - 1 |
7764 |
77 |
D50915.1 |
- |
1 - 1 |
7764 |
77 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
H65343.1 |
- |
1 |
|
|
Chr. 14 - 105932285 - 105932623 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AI220332.1 |
- |
1 |
|
|
Chr. 14 - 105932267 - 105932658 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AW449001.1 |
- |
1 |
|
|
Chr. 14 - 105932260 - 105932642 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 4537 |
MIR4537 |
|
-72.7kb
|
Sense |
Chr. - 105859484 - 105859553 on Build GRCh38 |
Downstream |
ADAM metallopeptidase domain 6 (pseudogene) |
ADAM6 |
C14orf96, tMDCIV |
37.1kb
|
Sense |
Chr. - 105969731 - 105972269 on Build GRCh38 |
View Full Product Details