Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
Chromosome Location
Chr.11: 113409595 - 113475279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016574.3 NP_057658.2 5 - 6 964 82
GenBank mRNA S69899.1 - 5 - 6 828 82

View Full Product Details