The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000795.3 | NP_000786.1 | 7 - 8 | 1374 | 62 |
NM_016574.3 | NP_057658.2 | 6 - 7 | 1287 | 62 | |
XM_017017296.1 | XP_016872785.1 | 7 - 8 | 1356 | 62 | |
GenBank mRNA | AB209832.1 | - | 6 - 7 | 1169 | 62 |
AB451480.1 | - | 5 - 6 | 1052 | 62 | |
AF176812.1 | - | 7 - 8 | 1154 | 62 | |
BC021195.2 | - | 7 - 8 | 1234 | 62 | |
M29066.1 | - | 6 - 7 | 1172 | 62 | |
M30625.1 | - | 7 - 8 | 1475 | 62 | |
S62137.1 | - | 6 - 7 | 1456 | 62 | |
X51362.1 | - | 7 - 8 | 1304 | 62 | |
X51645.1 | - | 6 - 7 | 1167 | 62 |