Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
Chromosome Location
Chr.10: 13100082 - 13138276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008211.1 NP_001008212.1 3 - 4 433 63
GenBank mRNA AF420371.1 - 3 - 4 290 63

View Full Product Details