Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
Chromosome Location
Chr.10: 13100082 - 13138276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008211.1 NP_001008212.1 5 - 6 829 108
NM_001008212.1 NP_001008213.1 4 - 5 760 108
NM_001008213.1 NP_001008214.1 5 - 6 814 108
NM_021980.4 NP_068815.2 3 - 4 608 108
GenBank mRNA AF061034.1 - 5 - 6 702 108
AF420371.1 - 5 - 6 686 108
AF420372.1 - 3 - 4 465 108
AF420373.1 - 4 - 5 617 108
AK055403.1 - 4 - 5 1049 108
AK303715.1 - 3 - 4 626 108
AK316171.1 - 3 - 4 648 108
BC013876.2 - 3 - 4 521 108
BC032762.1 - 5 - 6 734 108
KU178502.1 - 2 - 3 376 108
KU178503.1 - 2 - 3 376 108

View Full Product Details