Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
Chromosome Location
Chr.10: 13100082 - 13138276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008211.1 NP_001008212.1 7 - 8 1083 64
NM_001008212.1 NP_001008213.1 6 - 7 1014 64
NM_001008213.1 NP_001008214.1 7 - 8 1068 64
NM_021980.4 NP_068815.2 5 - 6 862 64
GenBank mRNA AF420371.1 - 7 - 8 940 64
AF420372.1 - 5 - 6 719 64
AF420373.1 - 6 - 7 871 64
AK055403.1 - 6 - 7 1303 64
AK303715.1 - 5 - 6 880 64
AK316171.1 - 5 - 6 902 64
BC013876.2 - 5 - 6 775 64
KU178502.1 - 4 - 5 630 64
KU178503.1 - 4 - 5 630 64

View Full Product Details