Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
Chromosome Location
Chr.10: 13100082 - 13138276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008211.1 NP_001008212.1 9 - 10 1336 80
NM_001008212.1 NP_001008213.1 8 - 9 1267 80
NM_001008213.1 NP_001008214.1 9 - 10 1321 80
NM_021980.4 NP_068815.2 7 - 8 1115 80
GenBank mRNA AF061034.1 - 9 - 10 1209 80
AF420371.1 - 9 - 10 1193 80
AF420372.1 - 7 - 8 972 80
AF420373.1 - 8 - 9 1124 80
AK055403.1 - 8 - 9 1556 80
AK303715.1 - 7 - 8 1133 80
AK316171.1 - 7 - 8 1155 80
BC013876.2 - 7 - 8 1028 80
BC032762.1 - 9 - 10 1223 80
KU178502.1 - 6 - 7 883 80

View Full Product Details