Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNK
Entrez Gene ID
85451
Gene Name
unkempt family zinc finger
Gene Aliases
UNKEMPT, ZC3H5, ZC3HDC5
Chromosome Location
Chr.17: 75784771 - 75825805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080419.2 NP_001073888.2 11 - 12 1589 68
XM_006722145.3 XP_006722208.1 14 - 15 5934 68
XM_006722148.2 XP_006722211.1 12 - 13 1730 68
XM_011525400.1 XP_011523702.1 12 - 13 1658 68
XM_011525401.2 XP_011523703.1 6 - 7 746 68
XM_017025247.1 XP_016880736.1 14 - 15 5970 68
XM_017025248.1 XP_016880737.1 13 - 14 5872 68
XM_017025249.1 XP_016880738.1 13 - 14 2015 68
XM_017025250.1 XP_016880739.1 12 - 13 1917 68
GenBank mRNA AB051540.1 - 11 - 12 1572 68
BC053362.1 - 11 - 12 1572 68

View Full Product Details