The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307922.1 | NP_001294851.1 | 1 - 2 | 285 | 85 |
NM_152682.3 | NP_689895.2 | 1 - 2 | 285 | 85 | |
GenBank mRNA | AK021702.1 | - | 1 - 2 | 208 | 85 |
AK315515.1 | - | 1 - 2 | 219 | 85 | |
BC017472.1 | - | 1 - 2 | 220 | 85 | |
BC067752.1 | - | 1 - 2 | 244 | 85 | |
BC107432.1 | - | 1 - 2 | 66 | 85 | |
DA250301.1 | - | 1 - 2 | 285 | 85 | |
DA766837.1 | - | 1 - 2 | 284 | 85 |