The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001961.1 | NP_001001961.1 | 1 - 1 | 551 | 82 |
GenBank mRNA | BC136949.1 | - | 1 - 1 | 594 | 82 |
BC136952.1 | - | 1 - 1 | 594 | 82 |