The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024334.2 | NP_077310.1 | 3 - 4 | 548 | 67 |
XM_017007176.1 | XP_016862665.1 | 4 - 5 | 670 | 67 | |
GenBank mRNA | AK027757.1 | - | 3 - 4 | 474 | 67 |
AK027877.1 | - | 3 - 4 | 412 | 67 | |
AK074073.1 | - | 1 - 2 | 84 | 67 | |
AK075010.1 | - | 3 - 4 | 399 | 67 | |
AK225589.1 | - | 3 - 4 | 471 | 67 | |
AL136916.1 | - | 3 - 4 | 442 | 67 | |
AY358625.1 | - | 3 - 4 | 311 | 67 | |
BC003125.1 | - | 3 - 4 | 443 | 67 | |
BC008054.2 | - | 1 - 2 | 38 | 67 | |
BC011719.2 | - | 3 - 4 | 443 | 67 | |
KU178769.1 | - | 2 - 3 | 294 | 67 | |
KU178770.1 | - | 2 - 3 | 294 | 67 |