Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr.3: 14124940 - 14143680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024334.2 NP_077310.1 4 - 5 641 73
XM_017007176.1 XP_016862665.1 5 - 6 763 73
GenBank mRNA AK027757.1 - 4 - 5 567 73
AK027877.1 - 4 - 5 505 73
AK074073.1 - 2 - 3 177 73
AK075010.1 - 4 - 5 492 73
AK225589.1 - 4 - 5 564 73
AL136916.1 - 4 - 5 535 73
AY358625.1 - 4 - 5 404 73
BC003125.1 - 4 - 5 536 73
BC008054.2 - 2 - 3 131 73
BC011719.2 - 4 - 5 536 73
KU178769.1 - 3 - 4 387 73
KU178770.1 - 3 - 4 387 73

View Full Product Details