The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024334.2 | NP_077310.1 | 8 - 9 | 966 | 66 |
XM_017007176.1 | XP_016862665.1 | 9 - 10 | 1088 | 66 | |
GenBank mRNA | AK027466.1 | - | 2 - 3 | 196 | 66 |
AK027757.1 | - | 8 - 9 | 892 | 66 | |
AK027827.1 | - | 3 - 4 | 249 | 66 | |
AK027877.1 | - | 8 - 9 | 830 | 66 | |
AK074073.1 | - | 6 - 7 | 502 | 66 | |
AK075010.1 | - | 8 - 9 | 817 | 66 | |
AK225589.1 | - | 8 - 9 | 889 | 66 | |
AL136916.1 | - | 8 - 9 | 860 | 66 | |
AY358625.1 | - | 8 - 9 | 729 | 66 | |
BC003125.1 | - | 8 - 9 | 861 | 66 | |
BC008054.2 | - | 6 - 7 | 456 | 66 | |
BC011719.2 | - | 8 - 9 | 861 | 66 | |
KU178769.1 | - | 7 - 8 | 712 | 66 | |
KU178770.1 | - | 7 - 8 | 712 | 66 |