Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr.3: 14124940 - 14143680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024334.2 NP_077310.1 9 - 10 1030 64
XM_017007176.1 XP_016862665.1 10 - 11 1152 64
GenBank mRNA AK027466.1 - 3 - 4 260 64
AK027757.1 - 9 - 10 956 64
AK027827.1 - 4 - 5 313 64
AK027877.1 - 9 - 10 894 64
AK074073.1 - 7 - 8 566 64
AK075010.1 - 9 - 10 881 64
AK225589.1 - 9 - 10 953 64
AL136916.1 - 9 - 10 924 64
AY358625.1 - 9 - 10 793 64
BC003125.1 - 9 - 10 925 64
BC008054.2 - 7 - 8 520 64
BC011719.2 - 9 - 10 925 64
KU178769.1 - 8 - 9 776 64

View Full Product Details