The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014439.3 | NP_055254.2 | 3 - 4 | 308 | 79 |
NM_173202.1 | NP_775294.1 | 2 - 3 | 245 | 79 | |
NM_173205.1 | NP_775297.1 | 2 - 3 | 188 | 79 | |
XM_011510963.2 | XP_011509265.1 | 4 - 5 | 2309 | 79 | |
GenBank mRNA | AB451333.1 | - | 2 - 3 | 203 | 79 |
AB451478.1 | - | 2 - 3 | 203 | 79 | |
AF167368.1 | - | 3 - 4 | 308 | 79 | |
AF200496.1 | - | 3 - 4 | 314 | 79 | |
AF201832.1 | - | 2 - 3 | 188 | 79 | |
AF251118.1 | - | 3 - 4 | 338 | 79 | |
AF251119.1 | - | 3 - 4 | 266 | 79 | |
AY071840.1 | - | 2 - 3 | 203 | 79 | |
BC020637.1 | - | 3 - 4 | 269 | 79 |