The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288990.1 | NP_001275919.1 | 2 - 3 | 415 | 74 |
NM_018430.3 | NP_060900.2 | 2 - 3 | 604 | 74 | |
XM_005256049.4 | XP_005256106.1 | 2 - 3 | 720 | 74 | |
XM_011523230.2 | XP_011521532.1 | 2 - 3 | 720 | 74 | |
XM_011523231.2 | XP_011521533.1 | 2 - 3 | 720 | 74 | |
XM_011523232.2 | XP_011521534.1 | 2 - 3 | 720 | 74 | |
XM_011523233.2 | XP_011521535.1 | 2 - 3 | 720 | 74 | |
XM_011523234.2 | XP_011521536.1 | 2 - 3 | 720 | 74 | |
XM_011523235.1 | XP_011521537.1 | 2 - 3 | 355 | 74 | |
XM_011523239.1 | XP_011521541.1 | 2 - 3 | 185 | 74 | |
GenBank mRNA | AK027245.1 | - | 2 - 3 | 322 | 74 |
AK301920.1 | - | 2 - 3 | 188 | 74 | |
BC111018.1 | - | 2 - 3 | 358 | 74 |