The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003906.4 | NP_003897.2 | 18 - 19 | 4115 | 126 |
XM_005261203.4 | XP_005261260.1 | 19 - 20 | 4691 | 126 | |
XM_005261204.4 | XP_005261261.1 | 19 - 20 | 4587 | 126 | |
XM_005261205.3 | XP_005261262.1 | 19 - 20 | 4203 | 126 | |
GenBank mRNA | AB005543.1 | - | 3 - 4 | 600 | 126 |
AB011144.1 | - | 18 - 19 | 3685 | 126 | |
AJ010089.1 | - | 18 - 19 | 4044 | 126 | |
AK126313.1 | - | 9 - 10 | 2297 | 126 | |
AK127239.1 | - | 8 - 9 | 2004 | 126 | |
AK304208.1 | - | 18 - 19 | 2974 | 126 | |
BC013285.2 | - | 3 - 4 | 401 | 126 | |
BC104958.1 | - | 18 - 19 | 4083 | 126 | |
BC104960.1 | - | 18 - 19 | 4083 | 126 |