Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location
Chr.18: 79069275 - 79378391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306085.1 NP_001293014.1 25 - 26 3034 70
NM_198531.4 NP_940933.3 25 - 26 3034 70
XM_011525963.2 XP_011524265.1 26 - 27 3278 70
XM_011525964.2 XP_011524266.1 25 - 26 2993 70
XM_011525965.2 XP_011524267.1 26 - 27 3274 70
XM_011525966.2 XP_011524268.1 25 - 26 3229 70
XM_011525971.2 XP_011524273.2 19 - 20 5336 70
XM_011525972.2 XP_011524274.1 20 - 21 2353 70
XM_011525973.2 XP_011524275.1 18 - 19 2127 70
XM_011525974.2 XP_011524276.1 18 - 19 2149 70
XM_017025726.1 XP_016881215.1 26 - 27 3278 70
XM_017025727.1 XP_016881216.1 26 - 27 3278 70
XM_017025728.1 XP_016881217.1 26 - 27 3200 70
XM_017025729.1 XP_016881218.1 26 - 27 3278 70
XM_017025730.1 XP_016881219.1 26 - 27 3200 70
XM_017025736.1 XP_016881225.1 26 - 27 3278 70
XM_017025737.1 XP_016881226.1 26 - 27 3278 70
XM_017025738.1 XP_016881227.1 22 - 23 2510 70
XM_017025739.1 XP_016881228.1 22 - 23 2528 70
XM_017025740.1 XP_016881229.1 21 - 22 2433 70
XM_017025741.1 XP_016881230.1 23 - 24 2773 70
GenBank mRNA AJ006268.1 - 4 - 5 70
AK127634.1 - 9 - 10 895 70
AL096735.1 - 2 - 3 79 70
BC125219.1 - 25 - 26 2912 70
BC125220.1 - 25 - 26 2912 70
BX537860.1 - 23 - 24 2417 70
BX648948.1 - 13 - 14 1609 70
U78978.1 - 5 - 6 469 70

View Full Product Details