The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031703.2 | NP_001026873.2 | 4 - 5 | 447 | 93 |
XM_005265241.3 | XP_005265298.1 | 3 - 4 | 373 | 93 | |
XM_011533851.2 | XP_011532153.1 | 4 - 5 | 449 | 93 | |
XM_011533852.2 | XP_011532154.1 | 4 - 5 | 785 | 93 | |
XM_011533853.2 | XP_011532155.1 | 4 - 5 | 426 | 93 | |
XM_011533854.2 | XP_011532156.1 | 3 - 4 | 350 | 93 | |
XM_011533856.2 | XP_011532158.1 | 3 - 4 | 405 | 93 | |
XM_011533857.2 | XP_011532159.1 | 3 - 4 | 575 | 93 | |
XM_017006669.1 | XP_016862158.1 | 4 - 5 | 449 | 93 | |
XM_017006670.1 | XP_016862159.1 | 4 - 5 | 718 | 93 | |
GenBank mRNA | AK000218.1 | - | 4 - 5 | 432 | 93 |
AK225721.1 | - | 4 - 5 | 429 | 93 | |
AK298202.1 | - | 3 - 4 | 382 | 93 | |
BC000623.2 | - | 4 - 5 | 351 | 93 | |
BC014204.2 | - | 4 - 5 | 351 | 93 | |
DQ787088.1 | - | 4 - 5 | 328 | 93 |