The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031703.2 | NP_001026873.2 | 5 - 6 | 640 | 86 |
XM_005265241.3 | XP_005265298.1 | 4 - 5 | 566 | 86 | |
XM_017006670.1 | XP_016862159.1 | 5 - 6 | 911 | 86 | |
GenBank mRNA | AK000218.1 | - | 5 - 6 | 625 | 86 |
AK225721.1 | - | 5 - 6 | 622 | 86 | |
AK298202.1 | - | 4 - 5 | 575 | 86 | |
BC000623.2 | - | 5 - 6 | 544 | 86 | |
BC014204.2 | - | 5 - 6 | 544 | 86 | |
DQ787088.1 | - | 5 - 6 | 521 | 86 |