Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 24 - 25 3920 124
XM_005265279.4 XP_005265336.1 26 - 27 4268 124
XM_005265280.3 XP_005265337.1 25 - 26 4085 124
XM_005265282.3 XP_005265339.1 26 - 27 4227 124
XM_005265283.3 XP_005265340.1 25 - 26 4082 124
XM_011533900.2 XP_011532202.1 28 - 29 4434 124
XM_011533902.2 XP_011532204.1 27 - 28 4366 124
XM_011533903.2 XP_011532205.1 27 - 28 4371 124
XM_017006725.1 XP_016862214.1 27 - 28 4341 124
XM_017006726.1 XP_016862215.1 28 - 29 4451 124
XM_017006727.1 XP_016862216.1 28 - 29 4396 124
XM_017006728.1 XP_016862217.1 27 - 28 4392 124
XM_017006729.1 XP_016862218.1 27 - 28 4797 124
XM_017006730.1 XP_016862219.1 28 - 29 4387 124
XM_017006731.1 XP_016862220.1 27 - 28 4370 124
XM_017006732.1 XP_016862221.1 27 - 28 4363 124
XM_017006733.1 XP_016862222.1 27 - 28 4760 124
XM_017006734.1 XP_016862223.1 27 - 28 4333 124
XM_017006735.1 XP_016862224.1 26 - 27 4754 124
XM_017006736.1 XP_016862225.1 26 - 27 4752 124
XM_017006737.1 XP_016862226.1 26 - 27 4674 124
XM_017006738.1 XP_016862227.1 26 - 27 4295 124
XM_017006739.1 XP_016862228.1 26 - 27 4671 124
XM_017006740.1 XP_016862229.1 26 - 27 4212 124
XM_017006741.1 XP_016862230.1 26 - 27 4303 124
XM_017006742.1 XP_016862231.1 26 - 27 4326 124
XM_017006743.1 XP_016862232.1 27 - 28 4725 124
XM_017006744.1 XP_016862233.1 26 - 27 4292 124
XM_017006745.1 XP_016862234.1 26 - 27 4646 124
XM_017006746.1 XP_016862235.1 26 - 27 4643 124
XM_017006747.1 XP_016862236.1 26 - 27 4705 124
XM_017006748.1 XP_016862237.1 25 - 26 4601 124
XM_017006749.1 XP_016862238.1 25 - 26 4167 124
XM_017006750.1 XP_016862239.1 24 - 25 4055 124
XM_017006751.1 XP_016862240.1 26 - 27 4228 124
XM_017006752.1 XP_016862241.1 27 - 28 4800 124
XM_017006753.1 XP_016862242.1 27 - 28 4799 124
XM_017006754.1 XP_016862243.1 27 - 28 4366 124
XM_017006755.1 XP_016862244.1 26 - 27 4303 124
XM_017006756.1 XP_016862245.1 26 - 27 4300 124
XM_017006757.1 XP_016862246.1 24 - 25 4026 124
XM_017006758.1 XP_016862247.1 24 - 25 4157 124
XM_017006759.1 XP_016862248.1 26 - 27 4228 124
XM_017006760.1 XP_016862249.1 27 - 28 4366 124
XM_017006761.1 XP_016862250.1 27 - 28 4375 124
XM_017006762.1 XP_016862251.1 27 - 28 4363 124
XM_017006763.1 XP_016862252.1 28 - 29 4425 124
XM_017006764.1 XP_016862253.1 26 - 27 4303 124
XM_017006765.1 XP_016862254.1 25 - 26 4182 124
XM_017006766.1 XP_016862255.1 26 - 27 4235 124
GenBank mRNA AF177387.1 - 23 - 24 3884 124
AF197569.1 - 24 - 25 3902 124
AF225870.1 - 22 - 23 3788 124
AF225871.1 - 22 - 23 3669 124
AF225872.1 - 23 - 24 3884 124
AY281068.1 - 23 - 24 3883 124
BC015323.1 - 1 - 2 175 124
BC089409.1 - 2 - 3 256 124
BC115009.1 - 23 - 24 3896 124
BC115010.1 - 23 - 24 3825 124
BC115011.1 - 23 - 24 3900 124
BC129934.1 - 24 - 25 3929 124
BC129935.1 - 24 - 25 3929 124

View Full Product Details