The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113239.2 | NP_001106710.1 | 3 - 4 | 1504 | 94 |
NM_022740.4 | NP_073577.3 | 3 - 4 | 1504 | 94 | |
XM_006715935.2 | XP_006715998.1 | 4 - 5 | 1493 | 94 | |
XM_011516077.2 | XP_011514379.1 | 3 - 4 | 2082 | 94 | |
XM_011516078.2 | XP_011514380.1 | 3 - 4 | 2083 | 94 | |
XM_011516079.2 | XP_011514381.1 | 3 - 4 | 2082 | 94 | |
XM_011516080.2 | XP_011514382.1 | 3 - 4 | 2083 | 94 | |
XM_011516081.2 | XP_011514383.1 | 3 - 4 | 1479 | 94 | |
XM_017012069.1 | XP_016867558.1 | 3 - 4 | 1585 | 94 | |
GenBank mRNA | AF207702.1 | - | 2 - 3 | 1212 | 94 |
AF208291.1 | - | 3 - 4 | 1341 | 94 | |
AF326592.1 | - | 3 - 4 | 1388 | 94 |