Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr.1: 166921121 - 167022214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286377.1 NP_001273306.1 8 - 9 1079 64
NM_001286378.1 NP_001273307.1 10 - 11 945 64
NM_032858.2 NP_116247.1 9 - 10 1172 64
XM_006711583.1 XP_006711646.1 10 - 11 976 64
XM_011510067.1 XP_011508369.1 9 - 10 1138 64
XM_011510068.1 XP_011508370.1 10 - 11 929 64
XM_017002602.1 XP_016858091.1 11 - 12 1313 64
XM_017002603.1 XP_016858092.1 10 - 11 975 64
GenBank mRNA AK027810.1 - 9 - 10 979 64
AK131066.1 - 9 - 10 982 64
AK302253.1 - 8 - 9 895 64
AL133073.1 - 8 - 9 736 64
BC028595.2 - 9 - 10 952 64
BC034310.1 - 9 - 10 1600 64
DQ076156.2 - 9 - 10 981 64
GU383113.1 - 8 - 9 888 64
GU383114.1 - 10 - 11 945 64

View Full Product Details