Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr.1: 166921121 - 167022214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286378.1 NP_001273307.1 3 - 4 259 66
NM_032858.2 NP_116247.1 2 - 3 486 66
XM_006711583.1 XP_006711646.1 3 - 4 290 66
XM_011510067.1 XP_011508369.1 2 - 3 461 66
XM_011510068.1 XP_011508370.1 3 - 4 252 66
XM_017002602.1 XP_016858091.1 4 - 5 627 66
XM_017002603.1 XP_016858092.1 3 - 4 289 66
GenBank mRNA AK027810.1 - 2 - 3 293 66
AK131066.1 - 2 - 3 296 66
AL133073.1 - 1 - 2 50 66
BC028595.2 - 2 - 3 266 66
DQ076156.2 - 2 - 3 295 66
GU383114.1 - 3 - 4 259 66

View Full Product Details