Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr.1: 166921121 - 167022214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286377.1 NP_001273306.1 2 - 3 493 111
NM_001286378.1 NP_001273307.1 4 - 5 359 111
NM_032858.2 NP_116247.1 3 - 4 586 111
XM_006711583.1 XP_006711646.1 4 - 5 390 111
XM_017002602.1 XP_016858091.1 5 - 6 727 111
XM_017002603.1 XP_016858092.1 4 - 5 389 111
GenBank mRNA AK027810.1 - 3 - 4 393 111
AK131066.1 - 3 - 4 396 111
AK302253.1 - 2 - 3 309 111
AL133073.1 - 2 - 3 150 111
BC028595.2 - 3 - 4 366 111
BC034310.1 - 2 - 3 871 111
DQ076156.2 - 3 - 4 395 111
GU383113.1 - 2 - 3 302 111
GU383114.1 - 4 - 5 359 111

View Full Product Details