Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORCS1
Entrez Gene ID
114815
Gene Name
sortilin related VPS10 domain containing receptor 1
Gene Aliases
hSorCS
Chromosome Location
Chr.10: 106573663 - 107181138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013031.2 NP_001013049.1 24 - 25 3442 100
NM_001206569.1 NP_001193498.1 24 - 25 3442 100
NM_001206570.1 NP_001193499.1 24 - 25 3442 100
NM_001206571.1 NP_001193500.1 24 - 25 3442 100
NM_001206572.1 NP_001193501.1 24 - 25 3442 100
NM_052918.4 NP_443150.3 24 - 25 3442 100
XM_011539199.2 XP_011537501.1 24 - 25 3299 100
XM_017015614.1 XP_016871103.1 24 - 25 3299 100
XM_017015615.1 XP_016871104.1 23 - 24 3167 100
XM_017015616.1 XP_016871105.1 24 - 25 2978 100
XM_017015617.1 XP_016871106.1 24 - 25 2777 100
XM_017015618.1 XP_016871107.1 16 - 17 1933 100
GenBank mRNA AB209142.1 - 24 - 25 3302 100
AF284756.1 - 24 - 25 3268 100
AK074581.1 - 2 - 3 148 100
AK123752.1 - 16 - 17 1996 100
AK126905.1 - 24 - 25 2806 100
AK289797.1 - 24 - 25 3443 100
AY099452.1 - 24 - 25 3268 100
AY099453.1 - 24 - 25 3268 100
BC131597.1 - 24 - 25 3268 100
EU281343.1 - 24 - 25 3268 100
EU281344.1 - 24 - 25 3268 100

View Full Product Details