The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002775.4 | NP_002766.1 | 4 - 5 | 1099 | 87 |
GenBank mRNA | AF070555.1 | - | 2 - 3 | 373 | 87 |
AK091944.1 | - | 6 - 7 | 874 | 87 | |
AK092476.1 | - | 5 - 6 | 880 | 87 | |
AK290089.1 | - | 4 - 5 | 590 | 87 | |
BC011352.1 | - | 5 - 6 | 866 | 87 | |
BC031082.1 | - | 4 - 5 | 1007 | 87 | |
D87258.1 | - | 4 - 5 | 1019 | 87 | |
Y07921.1 | - | 4 - 5 | 1009 | 87 |